Learn More
Description
eVol™ is the coupling of two precision devices—a digitally controlled electronic drive and an XCHANGE™ (patent pending) enabled analytical syringe. The XCHANGE™ analytical syringes can be easily and quickly changed, allowing them to be dedicated to individual liquids and methods, while preventing cross-contamination of reagents.
- Digitally controlled positive displacement dispensing system
- Programmable—enables user-independent liquid handling procedures
- Easily calibrated to ensure accurate dispensing
- Stores calibration factors for up to ten XCHANGE™ syringes
- Touch Wheel user interface
- Full color screen
Applications:
- Preparation of calibration standards
- Addition of internal standards
- Precise dispensing of non-aqueous liquids
- Sample dilution
Specifications
Specifications
| Needle Gauge | 25 G |
| Diameter (English) Inner Needle | 0.012 in. |
| Diameter (Metric) Inner Needle | 0.32 mm |
| Diameter (English) Outer Needle | 0.024 in. |
| Diameter (Metric) Outer Needle | 0.63 mm |
| For Use With (Application) | Preparation of calibration standards, addition of internal standards |
| Length (English) Needle | 1.96 in. |
| Product Type | eVol Analytical Syringe |
| Quantity | Ea. |
| Volume (Metric) | 500 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
