Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan™ BP21 (FFAP) GC Capillary Columns
SDP

Catalog No. SG054465 Shop All SGE Inc. Products
Click to view available options
Length (Metric):
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.5 μm
1 μm

A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids

  • Nitroterephthalic acid modified PEG polar phase
  • Ideal for low molecular weight acids
  • Applications include volatile free acids, fatty acid methyl esters (FAMEs), alcohols, aldehydes and ketones

Specifications

Functionality/Interaction Type Hydrogen Bonding or Dipole interactions
Diameter (Metric) Inner 0.25 mm
USP Type G25, G35
Stationary Phase Polyethylene Glycol
Max. Temperature 250°C
For Use With (Application) Acrylates Analysis, Fatty Acid Methyl Esters, Alcohol Analysis, Aldehydes Analysis, Acrylates, Ketones
Temperature Range 35°C to 240/250°C
Film Thickness 0.25 μm
Length (Metric) 30 m
Packing Material BP21 (FFAP)
Polarity Polar
Coating Fused Silica
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.