Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan™ GC Capillary Column Connectors: Replacement μ-Unions
SDP

Catalog No. p-4917460 Shop All SGE Inc. Products
Click to view available options
Connections:
0.1 - 0.32 mm and 0.53 mm
0.53 mm and 0.53 mm

Catalog No. SG073573

Supplier: Trajan™ 073573

SilTite μ-Union delivers an easy, leak-free installation for capillary columns without the use of any tools

  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches

Specifications

Type Replacement μ-Union
Connections 0.1 - 0.32 mm and 0.53 mm
For Use With (Equipment) GC Capillary Column Connectors
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.