Learn More
Description
- Allows plate mounting of union
- For use with fused silica
- Low dead volume
- High temperature applications
Specifications
Specifications
| Type | Bulkhead Union |
| Includes | Two ferrules, two nuts |
| Description | 0.25 mm restriction (at one end) |
| Material | Glass-lined Stainless Steel |
| Diameter (Metric) Inner | 0.4 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
