Learn More
SGE™ SilFlow™ GC 3 Port Splitter Kits
SDP
Catalog No. 03350233 Shop All SGE Inc. Products
Click to view available options
Diameter (Metric) Inner:
0.25/0.32 mm
0.53 mm
Port B and C 0.25/0.32 mm
Highly efficient and reliable micro fluidic platform to improve GC connectivity.
  • Kit includes a SilFlow microchannel device (MCD), SilTite FingerTite tool, mounting bracket, plus appropriate ferrules, nuts and blanking ferrules for set up
  • 1.1mm OD tubing not included
Specifications
Type Port Splitter
Diameter (Metric) Inner 0.25/0.32 mm
Includes SilFlow microchannel device (MCD), SilTite FingerTite tool, Mounting bracket, Appropriately sized ferrules, nuts and blanking ferrules for set up
For Use With SGE Silflow Microchannel Device
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.