Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dräger™ Short-Term Detector Tubes - Perchloroethylene
$166.83 - $480.39
| Catalog Number | Mfr. No. | Components Measured | Measurement Range | Humidity Range | No. of Strokes | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Components Measured | Measurement Range | Humidity Range | No. of Strokes | Price | Quantity & Availability | ||
|
17-985-283
|
Dräger™
8101501 |
Perchloroethylene 2/a | 2 to 300ppm | Less than25mg/L | 1/5 |
Pack of 10 for $166.83
|
|
||
|
19-046-631
|
Dräger™
8101551 |
Perchloroethylene 0.1/a | 0.5 to 4ppm | 30mg/L | 3/9 |
Pack of 10 for $480.39
|
|
||
Description
- Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
- Safety Equipment Institute (SEI) certified.
Specifications
| 10 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title