Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHP-2/PTPN11 Antibody (255509) [DyLight 350], Novus Biologicals™
SDP

Catalog No. FAB1894D Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1894D 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB1894D Supplier Novus Biologicals Supplier No. FAB1894D
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SHP-2/PTPN11 Monoclonal antibody specifically detects SHP-2/PTPN11 in Human,Mouse,Rat samples. It is validated for Western Blot,Gene Knock-Out

Specifications

Antigen SHP-2/PTPN11
Applications Western Blot, Gene Knock-Out
Classification Monoclonal
Clone 255509
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11
Host Species Rat
Immunogen E. coli-derived recombinant human SHP-2, Thr2-Arg593, Accession # NP_002825
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Lipid and Metabolism, Phospho Specific, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 5781
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.