Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIGIRR Antibody, Novus Biologicals™
SDP

Catalog No. NB10056890T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056890T 0.025 mg
NB10056890 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056890T Supplier Novus Biologicals Supplier No. NB10056890SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIGIRR Polyclonal specifically detects SIGIRR in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen SIGIRR
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5-1 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q6IA17
Gene Alias single Ig IL-1-related receptor, Single Ig IL-1R-related molecule, single immunoglobulin and toll-interleukin 1 receptor (TIR) domain, single immunoglobulin domain IL1R1 related, Single immunoglobulin domain-containing IL1R-related protein, TIR8MGC110992, Toll/interleukin-1 receptor 8
Gene Symbols SIGIRR
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 334-358 of human SIGIRIR.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59307
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.