Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Signal sequence receptor delta Antibody
SDP

Catalog No. NB10075620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10075620 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10075620 Supplier Novus Biologicals Supplier No. NB10075620

Chicken Polyclonal Antibody

Signal sequence receptor delta Polyclonal specifically detects Signal sequence receptor delta in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Signal sequence receptor delta
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS with No Preservative
Gene Accession No. P51571
Gene Alias signal sequence receptor, delta (translocon-associated protein delta), SSR-delta, translocon-associated protein delta, translocon-associated protein subunit delta, TRAP-delta, TRAPDSignal sequence receptor subunit delta
Gene Symbols SSR4
Host Species Chicken
Immunogen The immunogen for this antibody was a sequence from 61a.a. to 130a.a. from Signal sequence receptor delta(a 173a.a. long protein)
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6748
Test Specificity Signal sequence receptor delta
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.