Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega SignaTECT™ Protein Kinase Assay Systems

Catalog No. p-134670 Shop All Promega Corporation Products
Click to view available options
Target:
Calcium/Calmodulin

Contain proprietary SAM2 Biotin Capture Membrane for higher binding capacity and specificity

  • Produces lower backgrounds and higher signal-to-noise ratios as compared to traditional P81 phosphocellulose methods
  • Perforated and numbered membrane allows measurement of up to 96 kinase reactions; ideal for kinetic studies
  • Assay requires little “hands-on” manipulation and is complete in under an hour
  • Can be used in a variety of buffer/reaction conditions including cell extracts
  • Each assay system contains highly specific biotinylated peptide substrates for the appropriate kinase as well as necessary reaction components
  • PTK assay system also contains two proprietary peptide substrates

User must supply [γ-32P] ATP

U.S. Patents 6,066,462 and 6,348,310 and other patents pending.

Specifications

Target Calcium/Calmodulin
For Use With (Application) Can be used in a variety of Buffer or reaction conditions including cell extracts
Includes ATP, BSA, SAM2 Biotin capture membrane, Termination Buffer, CaM KII Biotinylated Peptide Substrate, reaction control, activation Buffer
Content And Storage -20°C
Product Type Protein Kinase Assay
Quantity 96 Reactions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.