Learn More
Cytiva Silicone Rubber Gaskets
Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
Upper Buffer Chamber
Description
- Made from silicone rubber
Specifications
Specifications
| For Use With (Application) | For use with SE 600 and SE 660 dual-cooled vertical IEF systems |
| For Use With (Equipment) | Upper Buffer Chamber |
| Type | Gasket |
| Includes | Slotted gasket |
| Material | Silicone Rubber |
| Quantity | 2/Pk. |