Learn More
Eisco™ Siltstone Specimen
SCI_ED
Catalog No. p-7076842 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Siltstone Specimen is a clastic rock made up of small, nearly microscopic grains of silt.
  • Clastic sedimentary rock made up of fine silt particles
  • For use in identifying depositional environments
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Clast size is indicative of low-energy environment
  • For clasts such as these to be deposited and lithified, they must be in environment with slowly moving or still water
Specifications
Product Type Sedimentary Rock, Siltstone
For Use With (Application) Identifying depositional environments
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.