Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Tamper Evident Screw Caps

Catalog No. p-4272603 Shop All Simport Scientific Products
Change view
Click to view available options
For Use With (Equipment):
30 mL Serum-standing Non-sterile Transport Tubes
5, 10 and 12 mL Serum-standing Non-sterile Transport Tubes
7 mL Serum-standing Non-sterile Transport Tubes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-040-250 7 mL Serum-standing Non-sterile Transport Tubes
22-040-249 5, 10 and 12 mL Serum-standing Non-sterile Transport Tubes
22-040-251 30 mL Serum-standing Non-sterile Transport Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-040-250 Supplier Simport™ Scientific Supplier No. T5527WTP
Only null left
Add to Cart
Add to Cart

Includes

Lip seal

Tamper Evident Screw Caps for all applications requiring tamper evident seal for utmost security and sample integrity.

  • Made of polyethylene
  • Features long skirt and super fast thread design
  • Can be removed or sealed with single turn
  • With leakproof seal

Specifications

Product Type Tamper-evident Screw Cap
Includes Lip seal
Material Polyethylene
Color White
For Use With (Equipment) 7 mL Serum-standing Non-sterile Transport Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.