Learn More
Scott Safety™ SKA-PAK™ Automatic Transfer Supplied Air Respirator
Catalog No. p-4625231
Click to view available options
Duration:
15 min.
3 min.
5 min.
Includes:
Dupont™ Kevlar™ aramid fiber harness, 4500psi carbon 15 min. cylinder, no facepiece, E-Z FLO Vibralert QD regulator, Schrader airline fitting
Nylon harness, 2216psi aluminum bottle, av-2000™ facepiece with Dupont™ Dupont™ Kevlar™ aramid fiber aramid fiber head harness, comfort seal, standard regulator and hansen fitting, vibralert™ tactile alarm system
Pressure demand supplied air respirator equipped with an automatic, hands-free air transfer capability.
  • Combination Type C positive pressure respirator
  • Automatic, hands-free transfer from external air source to cylinder in the event of primary air source interruption
  • Personal, tactile Vibralert alarm to alert users of transfer to cylinder air and enable prompt escape
  • Onboard cylinder gauge and 90% air remaining lighted alarm for added safety and awareness
  • E-Z Klip cylinder retention system for quick and easy cylinder changes
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NIOSH
Product Type Automatic Transfer Supplied Air Respirator
Duration 3 min.
For Use With (Application) Confined space, hazardous materials clean-up, chemical, petrochemical, oil and gas, and public utilities.
Includes Nylon harness, 2216psi aluminum bottle, av-2000™ facepiece with Dupont™ Dupont™ Kevlar™ aramid fiber aramid fiber head harness, comfort seal, standard regulator and hansen fitting, vibralert™ tactile alarm system
Model SAR422010331101
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.