Learn More
Description
Specifications
Specifications
| Antigen | SLC25A24 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Alias | APC1calcium-binding mitochondrial carrier protein SCaMC-1, calcium-binding transporter, DKFZp586G0123, MCSC1, Mitochondrial ATP-Mg/Pi carrier protein 1, mitochondrial ATP-Mg/Pi transporter, Mitochondrial Ca(2+)-dependent solute carrier protein 1, SCAMC1, SCAMC-1, short calcium-binding mitochondrial carrier 1, Small calcium-binding mitochondrial carrier protein 1, solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 24, Solute carrier family 25 member 24 |
| Gene Symbols | SLC25A24 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
