Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A25 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SLC25A25 |
---|---|
Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20-1:50 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SLC25A25 Polyclonal specifically detects SLC25A25 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC25A25 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
114789 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LRLVFKSLDKKNDGRIDAQEIMQSLRDLGVKISEQQAEKILKSMDKNGTMTIDWNEWRDYHLLHPVENIPEIILYWKHSTIFDVGENLT | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20-1:50 | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
APC3, calcium-binding mitochondrial carrier protein SCaMC-2, KIAA1896RP11-395P17.4, MCSC, MCSC3, MGC105138, MGC119514, MGC119515, MGC119516, MGC119517, Mitochondrial ATP-Mg/Pi carrier protein 3, Mitochondrial Ca(2+)-dependent solute carrier protein 3, mitochondrial Ca2+-dependent solute carrier, PCSCL, SCAMC2, SCAMC-2, short calcium-binding mitochondrial carrier 2, small calcium-binding mitochondrial carrier 2, Small calcium-binding mitochondrial carrier protein 2, solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 25, Solute carrier family 25 member 25, solute carrier family 25, member 25 | |
SLC25A25 | |
IgG | |
Affinity Purified | |
Specificity of human SLC25A25 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title