Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A48 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | SLC25A48 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179515
![]() |
Novus Biologicals
NBP179515 |
100 μL |
Each for $487.50
|
|
|||||
NBP17951520
![]() |
Novus Biologicals
NBP17951520UL |
20 μL | N/A | N/A | N/A | ||||
Description
SLC25A48 Polyclonal specifically detects SLC25A48 in Human samples. It is validated for Western Blot.Specifications
SLC25A48 | |
Polyclonal | |
Rabbit | |
Human | |
FLJ44862, HCC-down-regulated mitochondrial carrier protein, HDMCP, solute carrier family 25 member 48, solute carrier family 25, member 48 | |
SLC25A48 | |
IgG | |
31 kDa |
Western Blot | |
Unconjugated | |
RUO | |
BAC86705 | |
153328 | |
Synthetic peptide directed towards the c terminal of human LOC153328. Peptide sequence TAVGQLGNCHALLSPGGGQDTFRYSPNNSLLGTYSVPGPLPPQSHPFPMQ. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title