Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC38A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $648.50
Specifications
Antigen | SLC38A2 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SLC38A2 Polyclonal specifically detects SLC38A2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC38A2 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
54407 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MKKAEMGRFSISPDEDSSSYSSNSDFNYSYPTKQAALKSHYADVDPENQNFLLESNLGKKKYETEFHP | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
Amino acid transporter A2, ata2, ATA2System A transporter 1, KIAA1382System N amino acid transporter 2, pro1068, Protein 40-9-1, sat2, SAT2PRO1068, snat2, SNAT2amino acid transporter 2, sodium-coupled neutral amino acid transporter 2, Solute carrier family 38 member 2, solute carrier family 38, member 2, System A amino acid transporter 2 | |
SLC38A2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title