Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC7A5/LAT1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen SLC7A5/LAT1
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB124877
SDP
View Documents
Novus Biologicals
NBP309988100UL
100 μg
Each for $487.50
Only null left
Add to Cart
 
NB124878
SDP
View Documents
Novus Biologicals
NBP30998825UL
25 μg N/A N/A N/A
Description

Description

SLC7A5/LAT1 Polyclonal specifically detects SLC7A5/LAT1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

SLC7A5/LAT1
Western Blot
Unconjugated
Rabbit
Cancer
PBS buffer, 2% sucrose
8140
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
Mouse
4F2 LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1hLAT1, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, y+ system cationic amino acid transporter
The immunogen is a synthetic peptide directed towards the C terminal region of mouse SLC7A5/LAT1 (NP_003477.4). Peptide sequence IAVSFWKTPVECGIGFTIILSGLPVYFFGVWWKNKPKWLLQGIFSTTVLC
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.