Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slug Antibody, Novus Biologicals™
SDP

Catalog No. NBP227182SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227182SS 0.025 mg
NBP227182 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227182SS Supplier Novus Biologicals Supplier No. NBP227182SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Slug Polyclonal specifically detects Slug in Human, Mouse, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Slug
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS with 0.05% Sodium Azide
Gene Alias MGC10182, Neural crest transcription factor Slug, Protein snail homolog 2, slug homolog, zinc finger protein (chicken), SLUGH, SLUGzinc finger protein, snail 2, snail homolog 2 (Drosophila), WS2D, zinc finger protein SNAI2
Gene Symbols SNAI2
Host Species Rabbit
Immunogen Amino acids 82-110 of human SNAIL2 (SNAI2) were used as the immunogen.
Molecular Weight of Antigen 30 kDa
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuronal Stem Cell Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6591
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, horse and dog, 94% in mouse and 89% homologous in rat SNAIL2 (SNAI2).
Target Species Human, Mouse, Canine, Equine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.