Learn More
Thermo Scientific™ Smart-Vue™ 915MHz Wireless Monitoring Solution Kits (North America)
Increase security for lab samples with continous monitoring, datalogging and alarm notifications via Smart-Vue wireless solutions.
Specifications
Type Wireless Temperature Module with Sensor
Display Type LCD
Data Memory 3000 Readings
Alarm Types Telephone, E-mail, SMS, Fax, Audible, Visual, Printed Report
Software Type Smart-Vue Client, Smart-Vue Server
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Includes Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Includes Price Quantity & Availability  
SMARTKIT1
Accessories View Documents
Thermo Scientific™
SMARTKIT1
ULT Freezers Five calibrated wireless modules with PT100 sensors, client server software and 1 USB receiver N/A N/A N/A
SMARTKIT2A
Accessories View Documents
Thermo Scientific™
SMARTKIT2A
Refrigerators Five calibrated wireless modules with PT100 sensors N/A N/A N/A
Description
Description
Protect sample quality and facilitate regulatory compliance with Thermo Scientific™ Smart-Vue™ 915MHz Wireless Monitoring Solution Kits (North America). These solutions provide continuous monitoring and reporting of your laboratory equipment with immediate remote alarm notification and continuous data collection.
Sample Protection and Security
  • Permanently records all readings, alarms, and user acknowledgments
  • Protects work 24 hours a day, seven days a week with immediate remote alarm notification and continuous data collection
  • Audit trail traceability assists with conformance to 21 CFR part 11 and other regulatory requirements
  • Monitors temperatures ranging from -200° to +350°C, CO2 concentration, relative humidity (RH), differential pressure (clean rooms), 4-20 milliamps and dry contact state
Simplicity
  • Easy to install, use and maintain
  • No complicated wiring, allowing easy movement of laboratory equipment
  • Accessible anytime, anywhere via the internet with Smart-Vue client software
  • Quickly export reports to Microsoft™ Excel™, Word™ or Adobe™ PDF formats with just a click for auditing purposes
  • View multiple sensors
  • Alarm options: telephone, email, SMS text, fax, audible, visual and printed reports
Scalable
  • Single software solution, that is scalable and will grow with you, saving time and money
  • Easy to add new laboratory equipment to an existing Smart-Vue system
Complementary Services
  • Installation and validation services
  • Annual maintenance agreements
  • Calibration, inspection, battery replacement and software updates
  • Re-qualification (validation) as needed for cGxP compliance
Ordering Information:
Similar packaged solutions are available for cryopreservation systems, ovens, CO2 incubators, environmental chambers, water baths and other equipment. Smart-Vue modules and sensors are available for temperature, RH, CO2 concentration, differential pressure, 4-20mA output, ambient conditions, and alarm/relay dry contacts. Solutions vary by RF regions worldwide and are compatible with multiple brands and types of laboratory equipment. Please ask your Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements.
Compatible with:
Refrigerators, Freezers, LN2 Storage, CO2 Incubators, Clean Rooms, Storage Areas, Ovens
Recommended for:
For use in highly controlled environments, including pharmaceutical production, biotechnology, IVF clinics, clinical studies and diagnostic laboratories.
Note:
Some incubators have a decontamination feature; please note that the SMARTKIT3 is not designed to monitor temperatures over 120°C.
The purchase of Smart-Vue requires software installation, contact your sales representative for details.
Specifications
Specifications
Wireless Temperature Module with Sensor
3000 Readings
Smart-Vue Client, Smart-Vue Server
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
-200°C to +350°C
LCD
Telephone, E-mail, SMS, Fax, Audible, Visual, Printed Report
3.6 V Lithium
Differential Pressure Sensor, CO2 Sensor
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.