Learn More
Thermo Scientific™ Smart-Vue Pro Receivers
IT friendly LoRaWan provides easy set-up of a reliable, independent and secure wireless network
$1290.08 - $3430.09
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
SVHWRLGW001
|
Thermo Scientific™
SVPHWRLGW001 |
Smart-Vue Pro Indoor Receiver for LoRa Modules (Enable Ethernet or WiFi Connectivity) 915 Mhz with Universal Power Supply, External Antenna, Universal Power Supply, LoRaWAN Flyer | N/A | N/A | |||||
This product is not available for sales online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance. | |||||||||
11-731-063
|
Thermo Scientific™
SVPHWAPG915 |
Smart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with Antenna, Power Supply, 915 Mhz, no data/SIM |
Each for $1,290.08
|
|
|||||
11-731-060
|
Thermo Scientific™
SVPHWAPG915C |
Smart-Vue Pro MTC PRO Ethernet/4G LoRa Gateway Receiver with SIM card, data plan for up to 10 devices/1 year. Includes Antenna, Power Supply, 915 Mhz |
Each for $3,430.09
|
|
|||||
Description
LoRA is short for Long Range wireless Wide Area Network. It is a stand-alone dedicated network and increases connectivity range for IoT devices by 2 km in urban and 10 km in rural areas. This receiver pushes data to the Cloud by connecting to your local network via an Ethernet or Wi-Fi connection.
Long range wireless connectivity enables a single receiver to cover large areas, replacing the need for repeaters Dedicated wireless network requires only an internet connection, minimal IT support needed to set up. Support for up to 1000 data logging sensors per gatewaySpecifications
No Display |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.