Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMG1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB246829 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB246829 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB246829 Supplier Novus Biologicals Supplier No. NB10077278AF700
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMG1 Polyclonal antibody specifically detects SMG1 in Human samples. It is validated for Western Blot

Specifications

Antigen SMG1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ATXserine/threonine-protein kinase SMG1, EC 2.7.11, EC 2.7.11.1, hSMG-1,61E3.4, KIAA0421phosphatidylinositol 3-kinase-related protein kinase, Lambda/iota protein kinase C-interacting protein, Lambda-interacting protein, LIPPI-3-kinase-related kinase SMG-1, phosphatidylinositol 3-kinase-related kinase, SMG-1, SMG1 homolog, phosphatidylinositol 3-kinase-related kinase (C. elegans)
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1250 and 1300 of human PI-3-Kinase-related kinase SMG1 using the numbering given in entry NP_055907.3 (GeneID 23049).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 23049
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.