Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAP43 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | SNAP43 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18029620
![]() |
Novus Biologicals
NBP18029620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180296
![]() |
Novus Biologicals
NBP180296 |
100 μL |
Each for $487.50
|
|
|||||
Description
SNAP43 Polyclonal specifically detects SNAP43 in Human samples. It is validated for Western Blot.Specifications
SNAP43 | |
Polyclonal | |
Purified | |
RUO | |
NP_003073 | |
6617 | |
Synthetic peptide directed towards the C terminal of human SNAPC1. Peptide sequence GQGQVKATRKKEKKERLKPAGRKMSLRNKGNVQNIHKEDKPLSLSMPVIT. | |
Primary |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Proximal sequence element-binding transcription factor subunit gamma, PSE-binding factor subunit gamma, PTF subunit gamma, PTFgamma, small nuclear RNA activating complex, polypeptide 1, 43kD, small nuclear RNA activating complex, polypeptide 1, 43kDa, Small nuclear RNA-activating complex polypeptide 1, SNAP43SNAPc 43 kDa subunit, SNAPc subunit 1, snRNA-activating protein complex 43 kDa subunit, snRNA-activating protein complex subunit 1 | |
SNAPC1 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title