Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Potassium ATPase Beta 1 Antibody (464.8 (also known as 8A)), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 14 publications

$192.94 - $529.95

Specifications

Antigen Sodium Potassium ATPase Beta 1
Clone 464.8 (also known as 8A)
Dilution Western Blot 1:1000-1:5000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Paraffin reported in scientific literature
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB300147SS
SDP
View Documents
Novus Biologicals
NB300147SS
0.025 mL
Each for $192.94
Only null left
Add to Cart
 
NB300147
SDP
View Documents
Novus Biologicals
NB300147
0.1 mL
Each for $529.95
Only null left
Add to Cart
 
Description

Description

Sodium Potassium ATPase Beta 1 Monoclonal specifically detects Sodium Potassium ATPase Beta 1 in Human, Rat, Porcine, Bovine, Canine, Chicken, Hamster, Rabbit, Xenopus, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Sodium Potassium ATPase Beta 1
Western Blot 1:1000-1:5000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Paraffin reported in scientific literature
Monoclonal
Ascites
RUO
Human, Rat, Pig, Bovine, Canine, Chicken, Hamster, Rabbit, Xenopus species, Mouse (Negative)
Q9TT37
481
Purified Sodium Potassium ATPase Beta 1 from rabbit renal outer medulla. [UniProt# Q9TT37]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
464.8 (also known as 8A)
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction
Ascites with 0.1% Sodium Azide
adenosinetriphosphatase, ATP1B, ATPase, Na+/K+ transporting, beta 1 polypeptide, Beta 1-subunit of Na(+), K(+)-ATPase, MGC1798, Na, K-ATPase beta-1 polypeptide, sodium/potassium-dependent ATPase beta-1 subunit, Sodium/potassium-dependent ATPase subunit beta-1, sodium/potassium-transporting ATPase beta-1 chain, sodium/potassium-transporting ATPase subunit beta-1
ATP1B1
IgG2a κ
Unpurified
This is specific for Na,K-ATPase beta 1 subunit.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.