Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX2 Antibody (SOX2/1791), Novus Biologicals™
SDP

Catalog No. NB253407200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253407200 200 μg
NB25340720 20 μg
NB253407100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB253407200 Supplier Novus Biologicals Supplier No. NBP253407200UG

Mouse Monoclonal Antibody

SOX2 Monoclonal antibody specifically detects SOX2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, SDS-Page.

Specifications

Antigen SOX2
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SOX2/1791
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, ELISA 2 - 4 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, SDS-Page
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Gene Symbols SOX2
Host Species Mouse
Immunogen Recombinant human SOX2 protein fragment (aa176-305) (exact sequence is proprietary)
Molecular Weight of Antigen 34 kDa
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Core ESC Like Genes, Embryonic Stem Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6657
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.