Learn More
Bel Art™ Frigimat™ Junior Dry Ice Maker
Produces dry ice in flake or snow form
$6107.90
Specifications
For Use With (Application) Making Dry Ice
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
11-675-25
View Documents Promotion Details
Bel Art™
F388860000
Junior Dry Ice Maker
Case of 12 for $6,107.90
Only null left
Add to Cart
Description
Description
  • Screw nozzle directly to cylinder of compressed LCO2; cylinder must have a dip tube
  • When the cylinder valve is opened about 0.75 turn, LCO2 expands into the bag to create dry ice snow; The valve is then closed and the bag is ready to be removed and emptied
  • Secure bag to cone on the nozzle with the clamp
  • After emptying, the bag can be immediately reattached for next filling
  • Makes .77 kg (1.7 lb) of flakes per bag
Order Info
Cone, 5L x 2.312 in. dia. (127 x 59mm) at widest end. Bag, 6 dia. x 12 in. L (15 x 31cm).
Specifications
Specifications
Making Dry Ice
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.