Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cup Splitter for Siltek™ Liner

For high or low molecular weight compounds
$386.00 - $506.50
Specifications
Length (Metric) | 7.85 cm |
---|---|
Product Type | Siltek Cup Splitter Inlet Liner |
Quantity | 5/Pk. |
Diameter (Metric) Inner | 0.4 cm |
Diameter (Metric) Outer | 0.63 cm |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
06-717-813
![]() |
Restek™
20710213 |
Siltek Cup Splitter w/Wool |
Pack of 5 for $506.50
|
|
|||||
06-717-816
![]() |
Restek™
20710216 |
Siltek Cup Splitter w/CarboFrit |
Pack of 5 for $386.00
|
|
|||||
06-717-814
![]() |
Restek™
20710214 |
Siltek Cup Splitter |
Pack of 5 for $461.00
|
|
|||||
Description
- Increased sample residence time in the liner for homogeneous vaporization
- 4mm I.D.
- With Siltek™ deactivation
- Available with or without packing
Specifications
7.85 cm | |
5/Pk. | |
0.63 cm | |
Borosilicate Glass |
Siltek Cup Splitter Inlet Liner | |
0.4 cm | |
Agilent GCs Equipped with Split/Splitless Inlets |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title