Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 140333
GREENER_CHOICE

Catalog No. p-4816971
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent NK1 receptor antagonist

Potent NK1 receptor antagonist (Ki = 0.74 nM. IC50 = 1.6 nM). Inhibits substance P -invoked calcium mobilization and outward current (IC50 = 1.3 nM); blocks NK1-mediated nitric oxide-dependent vasodilation in vivo.

Chemical Identifiers

CAS 153050-21-6
Molecular Formula C37H45Cl3N2O2

Specifications

Quantity 10 mg
Formula Weight 656.12
Purity >97%
Molecular Weight (g/mol) 656.129
Chemical Name or Material SR 140333
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.