Learn More
Description
Specifications
Specifications
| Antigen | ssDNA |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | TNT-3 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Dilution | Simple Western, Flow Cytometry 5-10 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Gene Alias | Anti-DNA, Anti-ssDNA, Clone TNT3, Clone TNT-3, DNA DNA antibody TNT3, Necrotic marker, Single Stranded DNA, ssDNA TNT-3 monoclonal |
| Host Species | Mouse |
| Immunogen | Human Raji cell nuclei. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
