Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ssDNA Antibody (TNT-3), Novus Biologicals™
SDP

Catalog No. p-7200763 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229849 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229849 Supplier Novus Biologicals Supplier No. NBP229849
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ssDNA Monoclonal specifically detects ssDNA in All Species samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ssDNA
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TNT-3
Concentration 0.5mg/mL
Conjugate Unconjugated
Dilution Simple Western, Flow Cytometry 5-10 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias Anti-DNA, Anti-ssDNA, Clone TNT3, Clone TNT-3, DNA DNA antibody TNT3, Necrotic marker, Single Stranded DNA, ssDNA TNT-3 monoclonal
Host Species Mouse
Immunogen Human Raji cell nuclei.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with single stranded DNA (ssDNA). The antibody is useful for staining nucleated cells. The antibody can also be used to identify necrotic regions of tumors and damaged tissues. By immunohistochemistry, stains heterochromatic regions of cell nucleus.
Target Species All species
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.