Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ SUPELCOSIL™ ABZ+Plus HPLC Column, 5 μm Particle Size

Catalog No. p-7189748 Shop All MilliporeSigma Supelco Products
Click to view available options
Length (Metric):
15 cm
25 cm
Diameter (Metric):
3 mm
4 mm
4.6 mm
10 mm

Offer both high deactivation and unique selectivity.

SUPELCOSIL™ ABZ+Plus columns offer both high deactivation and unique selectivity. Deactivated silica particles of very narrow particle size distribution ensure high efficiency with low back pressure. After bonding and endcapping reactions, the ABZ+Plus phase effectively shields unreacted silanol groups on the silica, preventing them from interacting with most analytes, and provides symmetric peaks regardless of an analyte's functionality. The phase also allows you to use low ionic strength buffers without having to add an ion-suppressing modifier. ABZ+Plus enables you to use simple mobile phases when analyzing the most difficult compounds; acids, strongly basic compounds, and zwitterions. Suitable for L60 per USP.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 3 mm
Length (Metric) 25 cm
Particle Size 5 μm
Pore Size 120 Å
Surface Area 170 m2/g
Stationary Phase Amide, Alkyl
Product Line SUPELCOSIL
Matrix Amide, Alkyl Phase/Silica Gel, Spherical Particle Platform
Endcapped Yes
Phase Reversed Phase

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.