Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Survivin Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 341 publication
$243.50 - $487.50
Specifications
Antigen | Survivin |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Simple Western 1:25, Flow Cytometry reported in scientific literature (PMID 17875988; 33737139), ELISA reported in scientific literature (PMID 24102797), Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 12671708), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated, Dual RNAscope ISH-IHC |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB500201SS
![]() |
Novus Biologicals
NB500201SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB500201
![]() |
Novus Biologicals
NB500201 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
Survivin Polyclonal specifically detects Survivin in Human, Mouse, Rat, Canine, Feline, Guinea Pig, Hamster samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated, Dual RNAscope ISH-IHC.Specifications
Survivin | |
Western Blot 1:1000, Simple Western 1:25, Flow Cytometry reported in scientific literature (PMID 17875988; 33737139), ELISA reported in scientific literature (PMID 24102797), Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 12671708), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated, Dual RNAscope ISH-IHC | |
Polyclonal | |
Rabbit | |
Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells | |
PBS with 0.02% Sodium Azide | |
API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
BIRC5 | |
IgG | |
Affinity Purified | |
16 kDa |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Canine, Feline, Guinea Pig, Hamster | |
O15392 | |
332 | |
This Survivin Antibody was developed against full length recombinant human Survivin [UniProt# O15392] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title