Learn More
Invitrogen™ SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit
Description
Includes
100μL SuperScript III RT/Platinum Taq Mix; 2 x 1.25mL 2X SYBR Green Reaction Mix; 1mL Magnesium Sulfate (50mM); 100μL ROX Reference Dye (25μM); 300μL 20X Bovine Serum Albumin (1mg/mL)
The SuperScript™ III Platinum™ One-Step qRT-PCR Kit provides sensitive, specific detection with fluorogenic primers and probes, while the SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit offers the ease and convenience of SYBR™ Green I detection.
- Kit combines the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format
- The SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit offers the ease and convenience of SYBR™ Green I detection
- SuperScript™ III RT for higher temperature cDNA synthesis for greater success with difficult RNA secondary structure
- Platinum™ Taq DNA Polymerase with hot-start technology for improved specificity
- LUX™ Fluorogenic Primers or dual-labeled fluorogenic probes for superior detection performance
- SYBR™ Green I dye for easy and convenient detection
- Validated performance on multiple real-time instrument platforms, including rotor-based systems
- Store all components at -20°C (-80°C for long-term storage)
- ROX Reference Dye must be stored in the dark
Order Info
Shipping Conditions: Dry Ice
Specifications
Specifications
| Content And Storage | • 500 μL SuperScript™ III RT/Platinum™ Taq Mix • 12.5 mL 2X SYBR™ Green Reaction Mix • 2 × 1 mL Magnesium Sulfate (50 mM) • 500 μL ROX Reference Dye (25 μM) • 1.3 mL 20X Bovine Serum Albumin (1 mg/mL) Store all components at -20°C (-80°C for long-term storage). ROX Reference Dye must be stored in the dark. |
| Detection Method | SYBR |
| PCR Method | 1-step RT-qPCR |
| Polymerase | Taq DNA Polymerase |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ |
| Product Line | Platinum™, SYBR™, SuperScript™ |
| Product Type | One-Step qRT-PCR Kit |
| Quantity | 500 reactions |
| Shipping Condition | Dry Ice |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.