Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences SYBR™ Green-Based qPCR 2X Master Mix
SCI_ED

SYBR™ Green based qPCR 2X master mix is used for gene expression analysis, genotyping and mutation detection, pathogen detection, viral detection, absolute quantification. It has enhanced efficiency, specificity, sensitivity and compatible with nearly all real time PCR instruments.

$107.00 - $753.00

Specifications

Detection Method SYBR
Form Liquid
Reaction Speed Fast or Standard
Label or Dye ROX, Biotin, High Resolution Melting (HRM), SYBR Green, SYBR GreenER
Product Type qPCR Master Mix
View More Specs

Products 3
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
S03136
SCI_ED
View Documents
G-Biosciences
7861981
2000
Each for $753.00
Only null left
Add to Cart
 
S03137
SCI_ED
View Documents
G-Biosciences
7861980
500
Each for $242.00
Only null left
Add to Cart
 
S03138
SCI_ED
View Documents
G-Biosciences
7861979
100
Each for $107.00
Only null left
Add to Cart
 
Description

Description

  • SYBR™ Green based qPCR 2x master mix contains chemically modified hot start Taq DNA polymerase, SYBR™ Green I fluorescent dye, low ROX reference dye, dNTPs and optimized buffer with stabilizers.
  • This master mix is ideal for high-throughput real-time PCR screening and validation.
Specifications

Specifications

SYBR
Fast or Standard
qPCR Master Mix
Liquid
ROX, Biotin, High Resolution Melting (HRM), SYBR Green, SYBR GreenER
Gene Expression, Genotyping, Pathogen Detection, Virus Detection, Microbial Detection
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.