Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synapsin II Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $487.50

Specifications

Antigen Synapsin II
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:100
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018067
SDP
View Documents
Novus Biologicals
NB120132580.025MG
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB12013258
SDP
View Documents
Novus Biologicals
NB12013258
0.1 mg
Each for $487.50
Only null left
Add to Cart
 
Description

Description

Synapsin II Polyclonal specifically detects Synapsin II in Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

Synapsin II
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Neuroscience
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
6854
Synthetic peptide corresponding to a portion of rat Synapsin II. The sequence is completely conserved in mouse.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 1:100-1:2000, Immunoprecipitation 1:100
Polyclonal
Purified
RUO
Rat, Mouse
synapsin IISYNII, synapsin-2, SYNIIa, SYNIIb
SYN2
IgG
Protein A purified
This detects ∼74 and ∼54 kDa proteins, corresponding to the apparent molecular masses of synapsin IIa and IIb on SDS-PAGE immunoblots, in samples from mouse and rat origins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.