Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ SYTOX™ Dead Cell Stain Sampler Kit, for flow cytometry

Catalog No. 501121578
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 50 μL kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-1578 5 x 50 μL kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-112-1578 Supplier Molecular Probes™ Supplier No. S34862
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

SYTOX™ Dead Cell Stain Sampler Kit contains five nucleic acid stains that can be used to determine the viability of cells in flow cytometry assays.

  • Contains five nucleic acid stains: SYTOX™ Blue, SYTOX™ Green, SYTOX™ Orange, SYTOX™ AADvanced™, and SYTOX™ Red
  • Stains can be used to determine the viability of cells in flow cytometry assays
  • Each stain includes 50 tests to help identify the best stain to work in multi-color experiments
  • By flow cytometry, dead cells can be easily distinguished from live cells by the increased fluorescent signal due to dsDNA binding of SYTOX™ dyes
  • Stains have been optimized for use in cell concentrations ranging from 1 x 104 to 1x10 cells/mL
  • Validated in several cell types including Jurkat, ReH, CHO, Kg1a, 3T3, HeLa, HEK (primary cells), and lysed whole blood
  • Tested in several applications including apoptosis, cell cycle, cell proliferation, and immunophenotyping assays

Kit Includes:

  • SYTOX™ Blue Dead Cell Stain (S34857): For the 405 nm violet laser
  • SYTOX™ Green Dead Cell Stain (S34860): For the 488 nm blue laser with green emission
  • SYTOX™ AADvanced™ Dead Cell Stain (S10274, S10309): For the 488 nm blue laser with red emission
  • SYTOX™ Red Dead Cell Stain (S34854): For the 633/635 nm red laser
  • SYTOX™ Orange Dead Cell Stain (S34861): For the 532 nm green laser
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant
Color Blue, Green, Orange, AADvanced, Red
Content And Storage Contains 5 vials total - 1 vial each of SYTOX™ Blue, SYTOX™ Green, SYTOX™ Orange, SYTOX™ Red, SYTOX™ AADvanced™ dead cell stains in DMSO.

Store at ≤-20°C, desiccated and protect from light.
Description SYTOX™ Dead Cell Stain Sampler Kit, for flow cytometry
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dead Cell Stain Sampler Kit
Dye Type SYTOX™ Green, SYTOX™ Blue, SYTOX™ Orange, SYTOX™ AADvanced™, SYTOX™ Red
Emission 647/480/523/570/658
Excitation Wavelength Range 546/444/504/547/640nm
Form Solution
Format Tube(s)
Product Line SYTOX
Quantity 5 x 50 μL kit
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.