Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TrueMark™ Comprehensive Microbiota Control

The TrueMark Comprehensive Microbiota Control contains a linearized multi-target plasmid pool with microbial target sequences commonly used on Custom OpenArray panels. The control has targets relevant to many microbial profiling applications, including respiratory tract, urinary tract, vaginal microbiota, and antibiotic resistance research.

$2805.00 - $4910.00

Specifications

Detection Method Primer-probe
Format Tube
Product Line TrueMark
Shipping Condition Dry ice
View More Specs

Products 2
Catalog Number Mfr. No. Volume Price Quantity & Availability  
Catalog Number Mfr. No. Volume Price Quantity & Availability  
A50382
Encompass_Preferred
View Documents
Applied Biosystems™
A50382
1000 μL
Each for $2,805.00
Only null left
Add to Cart
 
A50383
Encompass_Preferred
View Documents
Applied Biosystems™
A50383
50 μL
Each for $4,910.00
Only null left
Edge
Add to Cart
 
Description

Description

The TrueMark Comprehensive Microbiota Control contains a linearized multi-target plasmid pool with microbial target sequences commonly used on Custom OpenArray panels. The control has targets relevant to many microbial profiling applications, including respiratory tract, urinary tract, vaginal microbiota, and antibiotic resistance research. The control can be included as a positive control for panel-specific amplification. It is available in two concentrations: 1 x 105 and 5 x 107 copies/μL. The higher concentration can be used to determine parameters necessary for analytical validation (e.g., limit of detection and Ct cut-off values).

The plasmid also contains target sequences for TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control, TaqMan Universal Extraction Control Organism (B. atrophaeus), and the human RNase P RPPH1 gene. During real-time PCR, the control can be used as a stand-alone sample to verify assay performance and to help with troubleshooting.

Note: This product will be updated on an annual basis to ensure coverage of the most popular microbial targets, and performance will be measured to ensure consistency between versions.

Version History:

Version 1 of this product was first available in April 2021 and contained 471 microbial assay targets.

Version 2 became available in October 2021 and contains an additional 83 microbial assay targets.

Version 3 became avalible in March 2023 and contains an additional 139 microbial and antibiotic resistant targets.

Specifications

Specifications

Primer-probe
Tube
TrueMark
Dry ice
Videos
Safety and Handling

Safety and Handling

ShelfLife : 2 years

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.