Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ PreAmp Master Mix (cDNA pre-amplification for qPCR)

Catalog No. p-4919403
Encompass_Preferred
Click to view available options
No. of Reactions:
200 Reactions
40 Reactions

TaqMan™ PreAmp Master Mix pre-amplifies small amounts of cDNA without introducing amplification bias to the sample.

TaqMan™ PreAmp Master Mix lets you: • Analyze mRNA from precious samples such as laser capture microdissections, needle biopsies, and formalin-fixed paraffin-embedded tissues (FFPE) • Stretch as little as 1 ng of cDNA into 200 real-time PCR reactions for gene expression analysis using TaqMan™ Gene Expression assays • Analyze up to 100 gene expression targets with minimal hands-on time Preserves Equilibrium of Targets Without Preamplification Bias: In the past, preamplification kits and techniques often resulted in uneven amplification of targets and incorrect or biased data (as shown by low correlation coefficients between preamplified and unamplified samples). TaqMan™ PreAmp Master Mix amplifies with no bias and provides extremely high correlation between amplified and unamplified cDNA. Easy Workflow: With TaqMan™ PreAmp Master Mix, preamplification setup takes only a few minutes. After a one-time pooling of the TaqMan™ assays , preamplification setup (mixing the pooled assays with cDNA sample and TaqMan™ PreAmp Master Mix) takes only five additional minutes. Cycling takes 1.5 hours using a GeneAmp™ 9700 thermal cycler. For Research Use Only. Not for use in diagnostic procedures.

Order Info

Guaranteed minimum shelf life is 60 days.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains one 5-mL bottle of 2X master mix.

Store at 2–8°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Detection Method Primer-probe
Format Bottle
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System
Product Line TaqMan™
Product Type PreAmp Master Mix
Quantity 200 reactions
Sufficient For 200 Reactions
For Use With (Application) Pre-amplification
No. of Reactions 200 Reactions
Sample Type cDNA
Volume 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.