Learn More
Promega TBK1 Kinase Enzyme System, 10microg
Catalog No. prv3991 Shop All Promega Corporation Products
Click to view available options
Quantity:
10 μg
Recombinant full-length human TBK1 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. TBK1, also known as NAK or NFkB-activating kinase, is an upstream protein kinase that can phosphorylate and activate the IkB kinases.
  • Easily Screen and Profile TBK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.