Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC-I 15
GREENER_CHOICE

Catalog No. p-4817059
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent α2β1 inhibitor; displays antithrombotic activity in vivo

TC-I 15 is a potent α2β1 integrin inhibitor (IC50 values for the inhibition of human platelet adhesion to type I collagen are 12 and 715 nM for platelets under static conditions and under flow, respectively). TC-I 15 displays selectivity for α2β1 over αvβ3, α5β1, α6β1 and αIIbβ3 at concentrations exceeding 1000 nM. TC-I 15 reduces collagen IV production in mesangial cells. Active in vivo; TC-I 15 prevents ferric chloride-induced clot formation in mice.

Chemical Identifiers

CAS 916734-43-5
Molecular Formula C23H28N4O6S2

Specifications

Quantity 10 mg
Formula Weight 520.62
Purity >98%
Molecular Weight (g/mol) 520.619
Chemical Name or Material TC-I 15
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.