Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TC21/R-Ras2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB342502 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342502 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342502 Supplier Novus Biologicals Supplier No. NBP281761MFV500
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TC21/R-Ras2 Polyclonal antibody specifically detects TC21/R-Ras2 in Human, Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen TC21/R-Ras2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias ras-related protein R-Ras2, related RAS viral (r-ras) oncogene homolog 2, TC21Ras-like protein TC21, Teratocarcinoma oncogene
Host Species Rabbit
Immunogen Rabbit polyclonal R-Ras2 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human R-Ras2. The immunogen is located within the last 50 amino acids of R-Ras2. Amino Acid Squence: PPSPEPTRKEKDKKG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 22800
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.