Learn More
Invitrogen™ TCAP Recombinant Rabbit Monoclonal Antibody (7E2Z4)
Rabbit Recombinant Monoclonal Antibody
$169.00 - $419.00
Specifications
Antigen | TCAP |
---|---|
Clone | 7E2Z4 |
Concentration | 1.1 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Western Blot |
Description
Immunogen sequence: MATSELSCEV SEENCERREA FWAEWKDLTL STRPEEGCSL HEEDTQRHET YHQQGQCQVL VQRSPWLMMR MGILGRGLQE YQLPYQRVLP LPIFTPAKMG ATKEEREDTP IQLQELLALE TALGGQCVDR QEVAEITKQL PPVVPVSKPG ALRRSLSRSM SQEAQRG.
Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. TCAP is a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in TCAP gene are associated with limb-girdle muscular dystrophy type 2G.Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. This gene encodes a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in this gene are associated with limb-girdle muscular dystrophy type 2G.Specifications
TCAP | |
1.1 mg/mL | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
O15273, O70548 | |
21393, 8557 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-167 of human TCAP (NP_003664.1). | |
Antibody |
7E2Z4 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human, Mouse | |
Tcap | |
19 kDa sarcomeric protein; AI503981; AJ223855; CMD1N; CMH25; LGMD2G; limb girdle muscular dystrophy 2G (autosomal recessive); Tcap; T-cap; TELE; telethonin; teneurin C-terminal associated peptide; titin cap protein; titin-cap; titin-cap (telethonin) | |
Tcap | |
Primary | |
Protein A |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.