Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCP1 alpha Antibody (23c (CTA-123)), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP20328A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20328A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20328A94 Supplier Novus Biologicals Supplier No. NBP250328AF594
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TCP1 alpha Monoclonal specifically detects TCP1 alpha in Mouse, Rat, Canine, Chicken, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TCP1 alpha
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 23c (CTA-123)
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha
Gene Symbols TCP1
Host Species Rat
Immunogen C-terminal half of the full length murine TCP protein
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6950
Test Specificity Does not recognize primate, xenopus or yeast TCP-1.
Target Species Mouse, Rat, Canine, Chicken, Hamster
Content And Storage Store at 4°C in the dark.
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.