Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Tenax™ GR Thermal Desorption Tubes

Catalog No. p-7202911
Click to view available options
Material:
Glass
Stainless Steel

Suitable for use with Air Monitoring

  • Fritted
  • Extends the range of Tenax™ TA
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Matrix Tenax GR Graphitized Porous Polymer
Product Line Tenax
For Use With (Application) Air Monitoring
Packaging Quantity 10/Pk.
Description Pre-conditioned
Diameter (English) Outer 1/4 in.
Overall Length (English) 3.5 in.
Material Glass
Type Thermal Desorption Tube

Tenax is a registered trademark of Buchem B.V.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.