Learn More
MilliporeSigma™ Supelco™ Tenax™ TA Thermal Desorption Tubes
Suitable for use with Air Monitoring
Manufacturer:  MilliporeSigma™ Supelco™ 29748U
 View more versions of this product
Catalog No. 11-100-5350
Add to cart
Description
Description
  • Fritted
  • Particle Size: 60/80 Mesh
Specifications
Specifications
Pre-conditioned/Sealed with Brass Endcaps
PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
3 1/2 in.
Thermal Desorption Tube
Tenax™ TA Porous Polymer (60/80)
10/Pk.
Air Monitoring
1/4 in.
Tenax™
Glass
EPA TO-1, IP-1 B
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Tenax is a registered trademark of Buchem B.V.