Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Teprotumumab Antibody (975547), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB345645 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345645 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345645 Supplier Novus Biologicals Supplier No. FAB96341MFV500
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Teprotumumab Monoclonal antibody specifically detects Teprotumumab in Broad Species Reactivity samples. It is validated for Flow Cytometry, Binding Inhibition

Specifications

Antigen Teprotumumab
Applications Flow Cytometry, Binding Inhibition Assay
Classification Monoclonal
Clone 975547
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Binding Inhibition
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Full-length Teprotumumab
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.