Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Ethyl Ether, 97%

Catalog No. ac403530250
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

Catalog No. AC403530250

Supplier: Thermo Scientific Chemicals 403530250

Only null left
Add to Cart

CAS: 637-92-3 | C6H14O | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-92-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00009225
InChI Key NUMQCACRALPSHD-UHFFFAOYSA-N
Synonym tert-butyl ethyl ether, ethyl tert-butyl ether, etbe, propane, 2-ethoxy-2-methyl, ethyl tert-butyl oxide, ether, tert-butyl ethyl, 2-methyl-2-ethoxypropane, ethyl t-butyl ether, tert-butylethylether, methyl-2-ethoxypropane
PubChem CID 12512
IUPAC Name 2-ethoxy-2-methylpropane
SMILES CCOC(C)(C)C

Specifications

Melting Point -94.0°C
Color Colorless
Density 0.7500g/mL
Boiling Point 71.0°C
Flash Point −19°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3746 to 1.3766
Linear Formula (CH3)3COC2H5
Quantity 25 g
Specific Gravity 0.75
Merck Index 15, 3829
Solubility Information Solubility in water: 1.2 g/100mL (20°C).
Formula Weight 102.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material tert-Butyl ethyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 211-309-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.