Learn More
Description
Specifications
Specifications
| Antigen | TFIIE beta |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | FE, General transcription factor IIE subunit 2, general transcription factor IIE, polypeptide 2 (beta subunit, 34kD), general transcription factor IIE, polypeptide 2, beta 34kDa, TF2E2TFIIE-beta, TFIIE-B, transcription initiation factor IIE subunit beta |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N terminal region of human TFIIE beta (NP_002086). Peptide sequence VEHGGSSGSKQNSDHSNGSFNLKALSGSSGYKFGVLAKIVNYMKTRHQRG |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
