Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ BetaBasic 4 CPF HPLC Columns
Click to view available options
:
Particle Size: 5μm; 150 x 1.0mm I.D.
Particle Size: 5μm; 150 x 2.1mm I.D.
Particle Size: 5μm; 150 x 3.0mm I.D.
Particle Size: 5μm; 150 x 4.6mm I.D.
Particle Size: 5μm; 250 x 1.0mm I.D.
Particle Size: 5μm; 250 x 2.1mm I.D.
Particle Size: 5μm; 250 x 3.0mm I.D.
Description
Thermo Scientific HPLC columns are manufactured under strict ISO 9001:2000 procedures, and every batch of silica and bonded phase is thoroughly tested for physical and chromatographic characteristics. Test reports are included with every column shipped.
- With Integral Column Plus Filter (CPF) System
- Fully deactivated Type B silica to ensure sharp peaks and excellent reproducibility
- Give less retention than C18 or C8 phases, and are a good choice for fast analyses including LC/MS of all types of compounds
- For basic compounds at high pH, which allows for a wide choice of buffer selection in LC/MS applications
- Stable bonding chemistry and excellent surface coverage gives the columns exceptional low and high pH stability

Specifications
Specifications
Diameter (Metric) | 2.1 mm |
Length (Metric) | 250 mm |
Particle Size | 5 μm |
Pore Size | 150 Å |
USP Type | L26 |
Stationary Phase | C4 |
Packing Material | Silica |
Carbon Load | 6% |
Column Type | Reversed Phase |
Endcapped | Yes |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction